Learn More
Tungsten oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Tungsten oxide is used as a pigment in ceramics and paints. It is used in various types of infrared absorbing coatings and foils. It is involved in the production of electrochromic windows, which is an electrically switchable glass that change light transmission properties with an applied voltage. It is a unique material utilized for various applications due their chromism, photocatalysis, and sensing capabilities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1500°C to 1600°C |
Color | Blue to Brown |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.99% metal basis |
Solubility Information | Soluble in hot alkalis. Insoluble in water and acids. |
Formula Weight | 230.25 |
Density | 7.16 |
Chemical Name or Material | Tungsten oxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z24K022 | Certificate of Analysis | 26/12/2024 | 089949.30,089949.18 |
Z04J038 | Certificate of Analysis | 15/12/2023 | 089949.30,089949.18 |
N06J068 | Certificate of Analysis | 24/10/2023 | 089949.30,089949.18 |
X25I061 | Certificate of Analysis | 04/11/2022 | 089949.30,089949.18 |
R02F009 | Certificate of Analysis | 08/12/2021 | 089949.30,089949.18 |
Safety and Handling

- Tungsten oxide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Tungsten oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.