missing translation for 'onlineSavingsMsg'
Learn More
Tungsten powder, -22 mesh, Puratronic™, 99.999% (metals basis), Thermo Scientific™ Product Code.: 15436005

Tungsten powder, -22 mesh, Puratronic™, 99.999% (metals basis), Thermo Scientific™

Product Code. 15436005
25 g
Click to view available options
Quantity:
2.5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15436005

Brand: Thermo Scientific Alfa Aesar 012973.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Color Gray
UN Number UN3089
Quantity 25 g
Percent Purity 99.999%
Grade Puratronic™
Mesh Size −22 Mesh
Form Powder
Assay (metals basis)
Chemical Name or Material Tungsten powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten powder, -22 mesh, Puratronic™, 99.999% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.