missing translation for 'onlineSavingsMsg'
Learn More
Tungsten powder, APS 12 micron, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11375878

Tungsten powder, APS 12 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11375878
2 kg
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11375878

Brand: Thermo Scientific Alfa Aesar 010401.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Color Gray
Appearance Light Gray powder
Assay Percent Range 99.9% (Metals basis)
Quantity 2 kg
Form Powder
Chemical Name or Material Tungsten

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP08H036 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number010401.09,010401.22,010401.A3,010401.36
Lot NumberU29A022 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number010401.A3
Lot NumberB09Q24 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010401.09,010401.22,010402.A3,010401.A3,010401.36
Lot NumberJ07X015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010401.09,010401.22,010402.A3,010401.A3,010401.36
Lot NumberH31P08 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number010401.09,010401.22,010402.A3,010401.A3,010401.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten powder, APS 12 micron, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.