missing translation for 'onlineSavingsMsg'
Learn More
Tungsten rod, 1.5mm (0.06 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11399877

Tungsten rod, 1.5mm (0.06 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11399877
30 cm
Click to view available options
Quantity:
30 cm
2 x 10 cm
5 x 30 cm
10 x 10 cm
This item is not returnable. View return policy

Product Code. 11399877

Brand: Thermo Scientific Alfa Aesar 042233.BV

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 30 cm
Percent Purity 99.95%
Weight ≈3.4g/10cm
Form Rod
Assay (metals basis)
Chemical Name or Material Tungsten rod, 1.5mm (0.06 in.) dia.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten rod, 1.5mm (0.06 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.