missing translation for 'onlineSavingsMsg'
Learn More

Tungsten rod, 5.0mm (0.20in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11450810
Click to view available options
Quantity:
10 cm
50 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 10 cm
Percent Purity 99.95%
Weight ≈3.7g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Tungsten rod, 5.0mm (0.20 in.) dia., Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'