Learn More
Tungsten sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Description
Tungsten sputtering target is used for Magnetron sputtering source. It is also used in electric filaments (e.g. in electric light bulbs),radiation shielding, military applications (e.g. armor and shells), as well as counter-balance materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Quantity | 1 Ea. |
| CAS | 7440-33-7 |
| Molecular Formula | W |
| MDL Number | MFCD00011461 |
| Synonym | wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder |
| Solubility Information | Insoluble in water. |
| InChI Key | WFKWXMTUELFFGS-UHFFFAOYSA-N |
| SMILES | [W] |
| IUPAC Name | tungsten |
| Molecular Weight (g/mol) | 183.84 |
| Afficher plus |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.