Learn More
Tungsten sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
Description
Tungsten sulfide is used as a component of catalysts for hydrotreating of crude oil, hydrodesulfurization and hydrodenitrification reactions. It finds application as solid-state secondary lithium battery cathodes and other electrochemical devices. It is found that it forms inorganic nanotubes, which is better reinforcing agents than carbon nanotubes. Tungsten sulfide addition to epoxy resin improves adhesion, fracture toughness and strain energy release rate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 1250°C (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.8% metal basis |
Molecular Formula | S2W |
Solubility Information | Slightly soluble in water. |
Formula Weight | 247.98 |
Odor | Odorless |
Density | 7.5 |
Show More |
RUO – Research Use Only