missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) chloride, 99%, 20 mesh

Product Code. 10392873 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10392873

Brand: Thermo Scientific Acros 200300100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13283-01-7 | Cl6W | 396.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13283-01-7
Molecular Formula Cl6W
Molecular Weight (g/mol) 396.57
MDL Number MFCD00011463
InChI Key KPGXUAIFQMJJFB-UHFFFAOYSA-H
Synonym tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
PubChem CID 83301
ChEBI CHEBI:37771
IUPAC Name hexachlorotungsten
SMILES Cl[W](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 347°C
Melting Point 275°C
Color Gray-Violet
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 99%
Linear Formula WCl6
Fieser 04,569; 05,742; 06,658; 14,346; 09,498; 10,444; 12,566; 13,343
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in cs2,chloroform,methyl alcohol,ether,,acetone,pyridine,glycerol,ligroin,ethanol,,carbon tetrachloride,phosphorus oxytrichloride,,benzene
Formula Weight 396.57
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Tungsten(VI) chloride, 20 Mesh
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.