missing translation for 'onlineSavingsMsg'
Learn More
Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10667202

Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10667202
10 g, Glass bottle
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10667202

Brand: Thermo Scientific Acros 221430100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13283-01-7
Molecular Formula Cl6W
Molecular Weight (g/mol) 396.57
MDL Number MFCD00011463
InChI Key KPGXUAIFQMJJFB-UHFFFAOYSA-H
Synonym tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungstenShow MoreShow Less
PubChem CID 83301
ChEBI CHEBI:37771
IUPAC Name hexachlorotungsten
SMILES Cl[W](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 347°C
Melting Point 275°C
Color Blue-Gray to Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9+%
Linear Formula WCl6
Fieser 04,569; 05,742; 06,658; 14,346
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in cs2,chloroform,methyl alcohol,ether,,acetone,pyridine,glycerol,ligroin,ethanol,,carbon tetrachloride,phosphorus oxytrichloride,,benzene
Formula Weight 396.57
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Tungsten(VI) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467467 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number221431000,221430100
Lot NumberA0463901 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number221431000,221430100
Lot NumberA0463901 Certificate TypeCertificate of Analysis Date21/06/2024 Catalog Number221431000,221430100
Lot NumberA0455708 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number221431000,221430100
Lot NumberA0459944 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog Number221431000,221430100
compliance-icons compliance-icons
Product Identifier
  • Tungsten(VI) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Tungsten chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.