missing translation for 'onlineSavingsMsg'
Learn More
Tungsten(VI) chloride, 99%, Thermo Scientific Chemicals Product Code.: 11301709

Tungsten(VI) chloride, 99%, Thermo Scientific Chemicals

Product Code. 11301709
5 g
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11301709

Brand: Thermo Scientific Alfa Aesar 012337.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a precursor for many tungsten compounds. It is used in tungsten plating, purification of tungsten, and olefin polymerization. It is also used as petroleum catalyst, and in intelligent glass. It can be used for deoxygenation of epoxides and as a reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13283-01-7
Molecular Formula Cl6W
Molecular Weight (g/mol) 396.54
MDL Number MFCD00011463
InChI Key KPGXUAIFQMJJFB-UHFFFAOYSA-H
Synonym tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
PubChem CID 83301
ChEBI CHEBI:37771
IUPAC Name hexachlorotungsten
SMILES Cl[W](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 346°C
Melting Point 275°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99%
Solubility Information Soluble in carbon disulfide,carbon tetrachloride and phosphorus oxychloride.
Formula Weight 396.57
Sensitivity Moisture sensitive
Density 3.52
Chemical Name or Material Tungsten(VI) Chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25K052 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number012337.30,012337.09,012337.18,012337.06
Lot NumberX23K007 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number012337.30,012337.09,012337.18,012337.06
Lot NumberX23K006 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number012337.30,012337.09,012337.18,012337.06
Lot NumberN06K007 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number012337.30,012337.09,012337.18,012337.06
Lot NumberN27J027 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012337.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten(VI) chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.