Learn More
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals
£86.40 - £278.00
Chemical Identifiers
CAS | 62571-53-3 |
---|---|
Molecular Formula | C12H30O6W |
Molecular Weight (g/mol) | 454.21 |
MDL Number | MFCD00156526 |
InChI Key | JHZFYDXXWXXVRM-UHFFFAOYSA-N |
Synonym | tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide |
PubChem CID | 13828235 |
IUPAC Name | ethanolate;tungsten |
SMILES | CCO[W](OCC)(OCC)(OCC)(OCC)OCC |
Description
Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62571-53-3 | |
454.21 | |
JHZFYDXXWXXVRM-UHFFFAOYSA-N | |
13828235 | |
CCO[W](OCC)(OCC)(OCC)(OCC)OCC |
C12H30O6W | |
MFCD00156526 | |
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide | |
ethanolate;tungsten |
Specifications
62571-53-3 | |
17°C (63°F) | |
C12H30O6W | |
MFCD00156526 | |
Moisture sensitive | |
Soluble in ethyl acetate,hydrocarbons and esters. | |
CCO[W](OCC)(OCC)(OCC)(OCC)OCC | |
454.21 | |
454.21 | |
5% w/v in ethanol | |
(metals basis) |
Brown to Yellow | |
Alcohol-like | |
W(OC2H5)6 | |
UN1170 | |
tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide | |
JHZFYDXXWXXVRM-UHFFFAOYSA-N | |
ethanolate;tungsten | |
13828235 | |
99.8% | |
Liquid | |
Tungsten(VI) ethoxide |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H225-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
For Manufacturing and Laboratory Use. Not for beverage use