missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) ethoxide, Thermo Scientific Chemicals

Product Code. 11338958
Click to view available options
Quantity:
1 g
5 g

Intermediate for sol-gel coatings

Intermediate for sol-gel coatingsTungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. Also, it acts as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62571-53-3
Molecular Formula C12H30O6W
Molecular Weight (g/mol) 454.21
MDL Number MFCD00156526
InChI Key JHZFYDXXWXXVRM-UHFFFAOYSA-N
Synonym tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide
PubChem CID 13828235
IUPAC Name ethanolate;tungsten
SMILES CCO[W](OCC)(OCC)(OCC)(OCC)OCC

Specifications

Melting Point 150°C
Color White to Brown
Linear Formula W(OC2H5)6
Quantity 1 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetateSoluble in ethyl acetate,hydrocarbons and esters.
Formula Weight 454.21
Physical Form Chunks and/or Powder
Chemical Name or Material Tungsten(VI) ethoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.