missing translation for 'onlineSavingsMsg'
Learn More
Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11381868

Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11381868
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11381868

Brand: Thermo Scientific Alfa Aesar 011828.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of Tungsten (VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-35-8
Molecular Formula O3W
Molecular Weight (g/mol) 231.837
MDL Number MFCD00011466
InChI Key ZNOKGRXACCSDPY-UHFFFAOYSA-N
Synonym tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
PubChem CID 14811
IUPAC Name trioxotungsten
SMILES O=[W](=O)=O

Specifications

Melting Point 1473°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Tungsten (VI) oxide is soluble in hot alkali solutions. Insoluble in water and acids. Slightly soluble in hydrofluoric acid.
Formula Weight 231.85
Density 7.16
Chemical Name or Material Tungsten(VI) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN25L010 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog Number011828.22,011828.36,011828.A3
Lot NumberN11L028 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog Number011828.22,011828.36,011828.A3
Lot NumberZ20K070 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number011828.22,011828.36,011828.A3
Lot NumberX03K038 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number011828.22,011828.36,011828.A3
Lot NumberR29K045 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011828.22,011828.36,011828.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.