missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) oxide, 99+%

Product Code. 10549823 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10549823

Brand: Thermo Scientific Acros 214215000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-35-8 | O3W | 231.837 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-35-8
MDL Number MFCD00011466
InChI Key ZNOKGRXACCSDPY-UHFFFAOYSA-N
Synonym tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa
PubChem CID 14811
IUPAC Name trioxotungsten
SMILES O=[W](=O)=O

Specifications

Melting Point 1470°C to 1475°C
Color Green-Yellow to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min
Molecular Formula O3W
Linear Formula WO3
Fieser 11,591
Merck Index 15, 9997
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in caustic alkalies,slightly soluble in acids
Molecular Weight (g/mol) 231.85
Formula Weight 231.85
Percent Purity ≥99%
Packaging Plastic bottle
Chemical Name or Material Tungsten(VI) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Tungsten(VI) oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.