missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Form: Approx. 2.13g/m

£47.84 - £193.97

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11395048
View Documents
Thermo Scientific Alfa Aesar
043995.G5
5 m
£47.84
5m
Please to purchase this item. Need a web account? Register with us today!
11305058
View Documents
Thermo Scientific Alfa Aesar
043995.H1
30 m
£193.97
30m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]

Specifications

Silver to Gray
Silver Gray wire
W
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Tungsten, Annealed
7440-33-7
99.95% (Metals basis)
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-143-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only