missing translation for 'onlineSavingsMsg'
Learn More
Tungsten wire, 1.25mm (0.049in) dia, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11386677

Tungsten wire, 1.25mm (0.049in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11386677
2 m
Click to view available options
Quantity:
10 m
2 m
This item is not returnable. View return policy

Product Code. 11386677

Brand: Thermo Scientific Alfa Aesar 000895.G2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powderShow MoreShow Less
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 2 m
Percent Purity 99.95%
Weight ≈23.7g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Tungsten wire, 1.25mm (0.049 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT13I035 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog Number000895.G2,000895.G6
Lot NumberK30Y049 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000895.G2,000895.G6
Lot NumberH19Q07 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000895.G2,000895.G6
Lot NumberA18L12 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000895.G2,000895.G6
Lot NumberK06R012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000895.G2,000895.G6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten wire, 1.25mm (0.049in) dia, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.