missing translation for 'onlineSavingsMsg'
Learn More
Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals Product Code.: 11304088

Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals

Product Code. 11304088
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11304088

Brand: Thermo Scientific Alfa Aesar 041375.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tungsten zirconium oxide is used as a super acid catalyst in the hydrocarbon isomerization process and esterification of waste acid oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16853-74-0
MDL Number MFCD00168096
Synonym Zirconium tungstate; Zirconium tungsten oxide

Specifications

Color Green to White
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 586.92
Percent Purity 99.7%
Odor Odorless
Density 5.10 g/mL
Assay (metals basis excluding Hf), Hf <0.3%
Chemical Name or Material Tungsten zirconium oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY04I010 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number041375.18,041375.30
Lot NumberJ21X036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041375.18,041375.30
Lot NumberL24G05 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041375.18,041375.30
Lot NumberA04R034 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041375.18,041375.30
Lot NumberD30U024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041375.18,041375.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.