missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungstic acid, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Green-Yellow to Yellow |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99% min |
Linear Formula | H2WO4 |
Merck Index | 15, 9998 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hf and alkalies |
Formula Weight | 249.86 |
Percent Purity | ≥99% |
Packaging | Glass bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'