missing translation for 'onlineSavingsMsg'
Learn More

Tungstic(VI) acid

Product Code. 11374929
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11374929

Brand: Thermo Scientific Alfa Aesar 082118.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-03-1 | H2O4W | 249.85 g/mol

Applied as flavoring agent and fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-03-1
Molecular Formula H2O4W
Molecular Weight (g/mol) 249.85
MDL Number MFCD00011336
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
Synonym tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g
PubChem CID 1152
ChEBI CHEBI:36272
IUPAC Name dihydroxy(dioxo)tungsten
SMILES O[W](=O)(=O)O

Specifications

Boiling Point 1473°C
Melting Point 100°C (decomposition)
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water and acid. Soluble in alkaline solution.
Formula Weight 249.86
Refractive Index 2.24
Density 5.5
Chemical Name or Material Tungstic(VI) acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.