missing translation for 'onlineSavingsMsg'
Learn More

Tungstosilicic acid hydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11334607
Click to view available options
Quantity:
10 g
50 g

Catalyst for organic synthesis, minerals separation, reagent for alkaloids.Tungstosilicic acid hydrate is used to prepare ethylacetate and acetic acid from ethylene. It is used in the preparation of silicotungstate butylmethylimidazolium bromide and charge transfer compounds with Keggin anions. It is also used in catalyst preparation, which is useful for catalytic liquid-phase oxidation of xylene. Further, it acts as a mediator in production of hydrogen through electrolysis of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-43-9
Molecular Formula H4O40SiW12
Molecular Weight (g/mol) 2878.16
MDL Number MFCD00036271
InChI Key HIKIIEGSHYIJCC-UHFFFAOYSA-K
Synonym tungstosilicic acid, tungstosilicic acid hydrate, p.a., tungstosilicic acid hydrate, purum p.a., tungstosilicic acid hydrate, reagent grade, tungstosilicic acid hydrate trace metals basis, tungstate 4-, m12-orthosilicato 4--ko:ko:ko:ko':ko':ko':ko:ko:ko:ko':ko':ko' tetracosa-m-oxododecaoxododeca-, hydrogen 1:4
PubChem CID 71307034
SMILES O[Si](O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)(O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O

Specifications

Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water.
IUPAC Name silicic acid;1,3,5-trioxa-2$l^{6},4$l^{6},6$l^{6}-tritungstacyclohexane 2,2,4,4,6,6-hexaoxide;hydrate
Formula Weight 2878.28 (Anhydrous)
Grade Reagent
Chemical Name or Material Tungstosilicic acid hydrate, Reagent Grade
compliance-icons
Product Identifier
  • Tungstosilicic acid hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tungstosilicic acidhydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'