Learn More
WTW™ Turbidimeter Cuvette
For automatic detection of the photLab™ turbidimeters and spectral photometers
Description
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.Specifications
Cuvette | |
3 Pack |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
WTW™ Turbidimeter Cuvette