Learn More
WTW™ Turbidity Standard
Turbidity standards AMCO Clear™ with the highest precision and long-term stability are N.I.S.T. traceable, lot certified and not hazardous to your health.
POA
Specifications
Type | Turbidity Standard |
---|---|
For Use With (Equipment) | Turb 555 IR |
Description | Calibration standards |
Description
- Long-term precision and stability
- No health hazards
- Easy to dispose of
- Adapted to the respective device optics in their particle composition
- Suitable for applications in the lower measuring range such as drinking water
Specifications
Turbidity Standard | |
Turb 555 IR | |
Calibration standards |