missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Turbo Cylindrical PTFE Stir Bars with Rare Earth Core

Cowie Technology™ Turbo Cylindrical PTFE Stir Bars with Rare Earth Core includes an assortment of useful Turbo Cylindrical Stirrer Bar sizes in a reusable container.

Brand:  Cowie Technology™ 001.3036.RE

Additional Details : Weight : 2.24000kg

Product Code. 10374209

  • £62.15 / 1 set
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Includes 15 Rare Earth Cylindrical Stirrer Bars
  • Constructed of PTFE with encapsulated Samarium-Cobalt magnet core
Specifications

Specifications

15 Rare Earth Cylindrical Stirrer Bars (Various Sizes)
195 x 95 x 30 mm
White
PTFE
Yes
195 mm
PTFE encapsulated Samarium-Cobalt magnet core
30 mm
1/Ea.
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers