Learn More
Thermo Scientific™ TurboFect™ Transfection Reagent
Deliver DNA plasmids and expression vectors into eukaryotic cells with this highly efficient, easy-to-use, non-immunogenic transfection reagent.
£64.50 - £964.00
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
12395613
|
Thermo Scientific™
R0533 |
200 μL |
£64.50
5 reactions |
In Stock
Log in to see stock. |
||||||
12331863
|
Thermo Scientific™
R0534 |
5 x 200 μL |
£283.00
5 reactions |
In Stock
Log in to see stock. |
||||||
15325016
|
Thermo Scientific™
R0531 |
1 mL |
£258.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15391506
|
Thermo Scientific™
R0532 |
5 x 1 mL |
£964.00
5mL |
In Stock
Log in to see stock. |
||||||
Description
The Thermo Scientific™ TurboFect in vitro Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery into eukaryotic cells. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells. Transfection can be performed in the presence or absence of serum. TurboFect Reagent demonstrates superior transfection efficiency and minimal toxicity when compared to other lipid-based or polymer-based transfection reagents.Highlights:
- High transfection efficiency of a wide variety of cell types
- No optimization required for most cell lines
- Excellent transfection efficiency in the presence or absence of serum
- Minimal cytotoxicity
- Ready-to-use – no need to reconstitute, dilute or manipulate
Recommended for:
Stable and transient transfection or co-transfection of DNA or RNA; Experiments with adherent or suspension cells, differentiated or undifferentiated cells; Gene expression studies and assays involving GFP or luciferase reporters
Specifications
Transfection Reagent |