missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Twaddle Hydrometers

Shot loaded Twaddle hydrometers

Brand:  Brannan™

 View more versions of this product

Product Code. 12839207

  • £28.16 / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For 0 to 24 Twaddell range subdivided 1.0TW. 158mm
Shot loaded, scale subdivision 1.0TW, accuracy ±1.0TW, 0.5TW ±0.5TW at 60/60°F. Calibrated at 15.6°C.
Specifications

Specifications

1Each
±1.0TW, 0.5TW ±0.5TW
UKAS
0 to 24 g/mL
Twaddle Hydrometer
1 g/mL
158 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brannan™ Twaddle Hydrometers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.