missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody (C06-3C8), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £372.00
Specifications
| Antigen | TWEAK/TNFSF12 |
|---|---|
| Clone | C06-3C8 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631719
|
Novus Biologicals
NBP3-11625-0.025mg |
0.025 mg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628106
|
Novus Biologicals
NBP3-11625-0.1mg |
0.1 mg |
£372.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for ELISASpecifications
| TWEAK/TNFSF12 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| PBS | |
| 8742 | |
| IgG1 | |
| Protein G purified |
| C06-3C8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| E. coli derived recombinant protein corresponding to aa 97-249 of human TWEAK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts