missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 blocking buffer, 1% in PBS (10X)

£50.05 - £133.67

Specifications

Product Type Blocking Buffer
Form Viscous Liquid
Concentration 1% in PBS (10X)
Content And Storage 2°C to 8°C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15464129
View Documents Promotion Details
Thermo Scientific Chemicals
J61544.K2
1 L
£69.70
1 L
Please to purchase this item. Need a web account? Register with us today!
15454129
View Documents Promotion Details
Thermo Scientific Chemicals
J61544.AP
500 mL
£50.05
500mL
Please to purchase this item. Need a web account? Register with us today!
15474129
missing translation for 'viewDocuments' Détails de la promotion
Thermo Scientific Chemicals
J61544.K3
2 L
£133.67
2 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunological, and eventually for incubation solutions of immunoreagents to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Spécification

Blocking Buffer
Viscous Liquid
1% in PBS (10X)
2°C to 8°C
Vidéos
Sécurité et traitement

Sécurité et traitement

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Certificats

RUO – Research Use Only