Learn More
Polysorbate 20, Fisher BioReagents™
Non-ionic detergent
£37.65 - £64.00
Specifications
Chemical Name or Material | Polysorbate 20 |
---|---|
Color | Amber |
CAS | 9005-64-5 |
Physical Form | Viscous Liquid |
Boiling Point | 100°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10113103
|
Fisher BioReagents
BP337-100 |
100 mL | Glass Bottle |
£37.65
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10485733
|
Fisher BioReagents
BP337-500 |
500 mL | HDPE Bottle |
£64.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Non-ionic detergent
This nonionic detergent is used in the isolation of water-soluble membrane proteins.Specifications
Polysorbate 20 | |
9005-64-5 | |
100°C | |
6 | |
Pass Test | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
3% max. | |
Polyoxyethylene-20-sorbitan Monolaurate, Polyoxyethylenesorbitan monolaurate | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 |
Amber | |
Viscous Liquid | |
1.100g/cm³ | |
96 to 108 | |
0.25% max. | |
400 mPa/s at 25°C | |
MFCD00165986 | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
443314 |
Safety and Handling

CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0
Recommended Storage : RT
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.