missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween™ 20, Pure, Fisher Chemical
Click to view available options
Quantity:
1 kg
Packaging:
Amber glass bottle
Specifications
Specifications
Color | Amber |
CAS | 9005-64-5 |
Physical Form | Very viscous Liquid |
Boiling Point | 100°C |
pH | 6 |
Quantity | 1 kg |
Packaging | Amber glass bottle |
Molecular Formula | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 |
Viscosity | 400 mPaS at 25°C |
MDL Number | MFCD00165986 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
15Minutes
0Seconds
21
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tween™ 20, Pure, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction