Learn More
Thermo Scientific Chemicals Tween 20 washing buffer, 1.0% in PBS (20X)
£109.00 - £193.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15405159
|
Thermo Scientific Chemicals
J62844.K3 |
2 L |
£193.00
2 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15495149
|
Thermo Scientific Chemicals
J62844.K2 |
1 L |
£109.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
It is used as wash buffers for ELISA, Western blot and other immunoassays as well as a blocking buffer for plate-based assays.