missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 washing buffer, 2% in PBS (20X)

Product Code. p-7082272
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 L
This item is not returnable. View return policy

Product Code. 15464029

Brand: Thermo Scientific Chemicals J61419.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PBS (Phosphate Buffered Saline) and Tween 20 are used as a wash buffer in immunolabeling techniques. Tween 20 is added to promote more effective washings, resulting in decreased non-specific background staining. Tween 20 is also known to promote reagent spreading in automated staining methodologies. 20X PBS Tween-20 is a space-saving stock solution that is ideal for preparing PBS-Tween (PBS-T) wash buffers for ELISA, Western and other immunoassays as well as a blocking buffer for plate-based assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer
Concentration 2% in PBS (20X)
Physical Form Liquid
Quantity 1 L

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.