missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 80, MP Biomedicals
43.33 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
100 mL
Specifications
CAS | 9005-65-6 |
Molecular Formula | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 |
MDL Number | MFCD00082107 |
Quantity | 100 mL |
InChI Key | HDTIFOGXOGLRCB-KTKRTIGZNA-N |
SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
IUPAC Name | 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
Molecular Weight (g/mol) | 604.82 |
Chemical Name or Material | Tween 80 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'