missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween™ 80 (Polysorbate), BAKER ANALYZED™, J.T. Baker™
159.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
500 mL
Specifications
CAS | 9005-65-6 |
Boiling Point | ∼100°C (1013 hPa) |
Melting Point | -21°C |
Density | 1.062 g/cm³ (20°C) |
Molecular Formula | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 |
MDL Number | MFCD00082107 |
Quantity | 500 mL |
Flash Point | >149°C |
InChI Key | HDTIFOGXOGLRCB-KTKRTIGZNA-N |
SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'