missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween™ 80, Pure, Fisher Chemical
Brand: Fisher Chemical T/4203/60
104.60 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
1kg, Amber glass bottle

Specifications
Amber | |
Liquid | |
1 kg | |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
9005-65-6 | |
6 | |
Amber glass bottle | |
MFCD00082107 | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
604.82 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction