missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
£66.90 - £112.45
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Autoclavable | Autoclavable |
Product Type | PCR Plate |
No. of Wells | 96 |
Product Code | Brand | Purity or Quality Grade | Volume (Metric) Well | Skirt Style | Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Purity or Quality Grade | Volume (Metric) Well | Skirt Style | Color | Price | Quantity & Availability | ||
10740920
|
Eppendorf™
0030132700 |
PCR Clean (Non-sterile) | 150 μL | Unskirted | White |
£112.45
Pack of 20 |
In Stock
Log in to see stock. |
||
10440378
|
Eppendorf™
0030132718 |
PCR Clean (Non-sterile) | 150 μL | Unskirted | Blue |
£110.00
Pack of 20 |
In Stock
Log in to see stock. |
||
15685367
|
Eppendorf™
0030129636 |
Forensic DNA Grade (Non-sterile) | 150 μL | Skirted | White |
£66.90
Pack of 10 Qty Price Break Available |
In Stock
Log in to see stock. |
||
15695367
|
Eppendorf™
0030129644 |
Forensic DNA Grade (Non-sterile) | 250 μL | Semi-skirted | White |
£66.90
Pack of 10 Qty Price Break Available |
In Stock
Log in to see stock. |
||
Description
- White wells for better reflection.
- High mechanical stability.
- Effective sealing by raised well rims.
- Skirted and semi-skirted frames.
- Optimal heat transfer due to reduced wall thickness.
- Autoclavable (121°C, 20 min.).
- Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
Specifications
Standard | |
Autoclavable | |
96 | |
6000 x g | |
White |
8 x 12 | |
PCR Plate | |
Non-sterile | |
Yes |