Learn More
Köttermann Two Door Tall Storage Cabinet
This Two Door Tall Storage Cabinet features a two hinged or sliding doors to protect stored equipment and utensils.
Brand:  Köttermann 301-00086
 View more versions of this product
Product Code. 17312099
  • £1595.00 / Each
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tall, slender design expands storage with a small footprint
  • Two doors protect stored items
  • Sliding and hinged door options available
Specifications
Specifications
White
Tall Storage
Hinged
Storage
None
None
No
No
516 x 900 x 1920 mm
Epoxy Coated
2
4
100.4 kg
Steel