missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Kel-F™ Fittings Assembled with PTFE FEP Tubing
Excellent for connecting syringes, equipment and other devices to Hamilton valves.
Brand: Hamilton™ 88918
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Manufactured from Kel-F™ PCTFE
- Tubing gauge of 12
- Tubing length of 304.8mm
Specifications
Cut End | |
Gauge: 12 | |
Kel-F |
1/4 in.-28 | |
304.8 mm |