missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Two-way Normally Closed Solenoid Pinch Valve
Prevent vale contamination
Brand: Masterflex™ 98302-02
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- Choose from two-way NC, two-way NO, or three-way pinch valves.
- Ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve.
- Tubing replacement is quick and easy.
- Suitable for pharmaceutical or sanitary applications.
- Mounting clip or mounting flange available separately for increased versatility.
Specifications
Pinch Valve for Tubing | |
1/16 in. | |
1/8 in. | |
15 psi |
Two-way normally closed solenoid pinch valve | |
0.0625 mm | |
1/16 in. ID, 1/8 in. OD |