Learn More
Thermo Scientific™ TX-750 4 x 750mL Swinging Bucket Rotor
Increase lab productivity with the 750 mL swinging bucket rotor, delivering maximum flexibility at 4700 rpm.
£124.00 - £2175.00
Specifications
Certifications/Compliance | Biocontainment certified by CAMR™, HPA, Porton Down, U.K. |
---|---|
Biocontainment | Yes |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15143169
|
Thermo Scientific™
75003180 |
TX-750 (Rotor body only) w/Slide Coat anti-friction finish |
£1650.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10683755
|
Thermo Scientific™
75003608 |
750mL Round Buckets (set of 4) |
£1490.00
4 sets |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606085
|
Thermo Scientific™
75003609 |
ClickSeal Biocontainment Lids for Round Buckets (set of 4) |
£381.00
Pack of 4 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11874451
|
Thermo Scientific™
75003610 |
Replacement O-rings for ClickSeal biocontainment lids (set of 4) |
£124.00
Pack of 4 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10247574
|
Thermo Scientific™
75003614 |
250mL Rectangular Buckets (set of 4) |
£2175.00
4 sets |
Estimated Shipment: 20-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10389874
|
Thermo Scientific™
75003615 |
ClickSeal Biocontainment Lids for Rectangular Buckets (set of 4) |
£385.00
Pack of 4 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10718975
|
Thermo Scientific™
75003617 |
Microplate/Flask Carriers, Set of 4 |
£2015.00
Pack of 4 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11848143
|
Thermo Scientific™
75003795 |
Microplate/Flask Carriers, Set of 2 |
£1040.00
2 sets |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Increase lab productivity with the Thermo Scientific™ TX-750 4 x 750mL Swinging Bucket Rotor, delivering maximum flexibility at 4700rpm.
Ordering Information:Buckets, lids and microplate carriers are sold separately; order TX-750 rotor body (75003180) and components to complete assembly. For replacement O-rings for lids (set of four) order 75003610. Tube adapters are also available separately. Contact your sales representative for information.
Includes:
Rotor manual
Compatible with:
Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Specifications
Biocontainment certified by CAMR™, HPA, Porton Down, U.K. | |
Yes |