Learn More
DuPont™ Tychem 2000 C
Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
Brand: DuPont™ D13395699
402.54 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Code : NEW
Additional Details : Weight : 10.17000kg
Description
- Hooded coverall with elastic face opening
- Adhesive chin flap for tight fit around respirator
- Stitched and over-taped seams
- Souble self-adhesive zipper flap
- Elastic cuffs, ankles and waist
- Thumb loops prevent sleeves from riding up
- 100% particle tight fabric
- Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
- Antistatic treatment (inside)
Specifications
Hooded Coveralls | |
Tychem | |
Elastic | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Polymeric barrier coating | |
For Maintenance and Dismantling Jobs in the Nuclear Industry |
X-Large | |
Yellow | |
Elastic | |
Collared | |
Double Zipper with Self-adhesive Flap | |
Category III, Type 3-B, 4-B, 5-B and 6-B | |
EN 14126, EN 1073-2, EN 1149-5 |