missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem 2000 C

Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).

Brand:  DuPont™ D13395560

402.54 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11753093

  • £503.00 / Pack of 25
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hooded coverall with elastic face opening
  • Adhesive chin flap for tight fit around respirator
  • Stitched and over-taped seams
  • Souble self-adhesive zipper flap
  • Elastic cuffs, ankles and waist
  • Thumb loops prevent sleeves from riding up
  • 100% particle tight fabric
  • Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
  • Antistatic treatment (inside)

Order Info

dimSize

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hooded Coveralls
Tychem
Elastic
Thumb Loop
Elastic
Stitched, Taped
Chemicals
Tyvek™ C Fabric
Tychem™ C is used for splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
2X-Large
Yellow
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
EN 14126, EN 1073-2, EN 1149-5
116 to 124 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers