Learn More
DuPont™ Tychem 2000 C
Bright yellow color provides high visibility. Lightweight and durable fabric and a polymeric barrier coating offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
Brand: DuPont™ D13494969
16.10 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Code : 94
Additional Details : Weight : 0.32400kg
Description
- Hooded coverall with elastic face opening
- Adhesive chin flap for tight fit around respirator
- Stitched and over-taped seams
- Souble self-adhesive zipper flap
- Elastic cuffs, ankles and waist
- Thumb loops prevent sleeves from riding up
- 100% particle tight fabric
- Excellent barrier against a broad range of concentrated inorganic chemicals and biological hazards
- Antistatic treatment (inside)
Specifications
Hooded Coveralls | |
Tychem | |
Elastic | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Chemicals | |
Tyvek™ C Fabric | |
Tychem™ C is used for splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing |
Medium | |
Yellow | |
Elastic | |
Hooded | |
Double Zipper with Self-adhesive Flap | |
Category III, Type 3-B, 4-B, 5-B and 6-B | |
EN 14126, EN 1073-2, EN 1149-5 | |
92 to 100 cm |