missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 S with Socks
GREENER_CHOICE

Product Code. 15939416 Shop All DuPont Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
This item is not returnable. View return policy
MTO: Made to order terms & conditions apply. Working in Ex-Zones: Please take this into account for your risk-assessment that the attached socks may isolate the wearer. There is the possibility that the garment and wearer cannot by grounded via the shoes and other measures for grounding the garment and the wearer are required.The garment does not protect against ionizing radiation.This garment and/or fabric are not flame resistant and should not be used around heat, open flame, sparks or in potentially flammable environments.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15939416

Brand: DuPont™ D15193524

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
MTO: Made to order terms & conditions apply. Working in Ex-Zones: Please take this into account for your risk-assessment that the attached socks may isolate the wearer. There is the possibility that the garment and wearer cannot by grounded via the shoes and other measures for grounding the garment and the wearer are required.The garment does not protect against ionizing radiation.This garment and/or fabric are not flame resistant and should not be used around heat, open flame, sparks or in potentially flammable environments.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tychem™ 4000 garments are made from a lightweight, supple and durable fabric. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.

  • DuPont™ Tychem™ 4000 S with socks. Hooded coverall with integrated socks and additional knee-length boot flaps. Available in white, in sizes SM to 3X. Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up. Self-adhesive chin flap for tight seal of suit to face-mask.
    Tychem™ 4000 garments are made from a lightweight and durable fabric (<800g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek™ substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
    Tychem™ 4000 S is ideal for chemical handling, environmental clean-up operations and emergency response. It is suitable for use in a variety of industries, including oil and gas, chemical engineering, and for use by hazardous material response teams and other emergency services.
  • For enhanced liquid protection, taping of outer cuff to glove is recommended. A double cuff is not a substitute for supplementary taping.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Elastic
Closure Type Double Zipper Front with Double Flaps
Seam Type Stitched, Taped
Sterility Non-sterile
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B
Resistance Chemicals, Liquid, Particle, Anti-static, Abrasion
Certifications/Compliance Certified according to Regulation (EU) 2016/425Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-BEN 14126 (barrier to infective agents), EN 1073-2 (protection againstradioactive contamination)Antistatic treatment (EN 1149-5) - on inside;
Description Tychem 4000 S with socks
No. of Pockets None
Size (English) Chest 39-43 in.
Size (Metric) Chest 100 to 108 cm
For Use With (Application) Chemical handling, environmental clean-up operations and emergency response.
Height (Metric) 174 to 182 cm
Weight (English) 124 g/m2
Quantity 15/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.