missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 4000 S with Socks
Shop All DuPont Products

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
Description
- DuPont™ Tychem™ 4000 S with socks. Hooded coverall with integrated socks and additional knee-length boot flaps. Available in white, in sizes SM to 3X. Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up. Self-adhesive chin flap for tight seal of suit to face-mask.
Tychem™ 4000 garments are made from a lightweight and durable fabric (<800g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek™ substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
Tychem™ 4000 S is ideal for chemical handling, environmental clean-up operations and emergency response. It is suitable for use in a variety of industries, including oil and gas, chemical engineering, and for use by hazardous material response teams and other emergency services. - For enhanced liquid protection, taping of outer cuff to glove is recommended. A double cuff is not a substitute for supplementary taping.
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | X-Large |
| Gender | Unisex |
| Material | Tychem |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Closure Type | Double Zipper Front with Double Flaps |
| Seam Type | Stitched, Taped |
| Sterility | Non-sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction