missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 AL TF630 WG
144.07 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Use promo code "22918" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Ventilated coverall available in sizes small to 3X.
- Attached non-dissipative under gloves.
- Integrated dissipative socks with boot flap.
- Gray.
- The protective system made of Tychem™ 6000 AL TF630 WG and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates.
- The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
- The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip.
- In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
- Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate.
- They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray |
For Use With (Application) | Protecting from Contaminents |
Hood Type | Fully Encapsulated |
Material | Tychem |
Size | Small |
Closure Type | Horizontal Zipper with Double Flap Hook and Loop |
Disposable | Single-use |
Gender | Unisex |
Latex Free | Yes |
No. of Pockets | None |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'