missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tychem™ 6000 F FaceSeal Product Code.: 16388555
Estimated Shipment: 22-05-2025
to see stock.
Add to basket

DuPont™ Tychem™ 6000 F FaceSeal

Product Code. 16388555 Shop All DuPont Products
5X-Large
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16388555

Brand: DuPont™ D15490762

Estimated Shipment: 22-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tychem™ 6000 F FaceSeal is a hooded coverall that includes a rubber seal for a tight fit.

  • Made from a lightweight and durable fabric (< 500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate
  • Offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Back entry with zipper, double zipper flap with snaps and hook and loop closure
  • Attached undergloves
  • Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Size 5X-Large
Ankle Style Attached socks
Closure Type Rear-entry zipper with double flap, snaps and hook-and-loop seal
Cuff Style Attached gloves
Disposable Single-use
Gender Unisex
Neck Style Hooded
Quantity 1/Ea.
Seam Type Stitched, Taped
Sterility Non-sterile
Waist Style Elastic
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tychem™ 6000 F FaceSeal >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.