missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Plus
287.23 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- DuPont™ Tychem™ 6000 F Plus. Hooded coverall available in grey. Elasticated face, wrists, waist and ankles for an optimal fit. Double cuffs, knitted inner cuffs, thumb loops to prevent sleeves from riding up. Front entry with double zipper, double flap and adhesive tape at outer flap.
Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Tychem™ 6000 F garment are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
- Certified according to tradeulation (EU) 2016/425
- Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) - on inside
- Stitched and over-taped seams with barrier tape for protection and strength
- Front closure with double zipper, double flap, outer flap with adhesive tape
- Double cuffs, thumb loops, knitted inner cuffs
Specifications
Product Type | Chemical Protective Clothing |
Size | Large |
Gender | Unisex |
Material | Tychem |
Color | Gray |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Thumb Loop, Elastic |
Neck Style | Hooded |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'