Learn More
DuPont™ Tychem™ 6000F Gown
553.39 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Use promo code "22918" to get your promotional price.
Click to view available options
Size:
Medium
2X-Large
Description
- Elasticated wrists
- Knitted cuff (inside) for better comfort
- Mandarin collar for improved neck and throat area closure
- Offers excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | 2X-Large |
Color | White |
Cuff Style | Elasticated |