Learn More
DuPont™ Tychem™ 6000F Hooded Coverall with FaceSeal Product Code.: 16335035
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
DuPont™ Tychem™ 6000F Hooded Coverall with FaceSeal
Product Code. 16335035 Shop All DuPont Products
Type: Coverall
Click to view available options
Type: Coverall
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16335035
Brand: DuPont™ D15490543
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Hooded coverall with rubber seal on hood.
  • Elasticated waist for an optimal fit
  • Double cuffs with attached undergloves
  • Dissipative socks with boot flap and back entry for enhanced wearer protection
  • Lightweight and durable fabric
  • Offers excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (EU) 2016/425, EN 14126, EN 1149-5
Color Gray
Size X-Large
Closure Type Zipper
Includes Attached dissipative socks and boot flaps
Product Type Coverall
Seam Type Stitched and Taped
Product Content Correction
Product Title
DuPont™ Tychem™ 6000F Hooded Coverall with FaceSeal >