missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Antistatic treatment
- Wide elastics at cuffs and upper arm
- Utilizes the strength of Tyvek and a polymeric barrier coating to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
- Used for splash or pressurized splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing

Specifications
Specifications
Size | Universal |
Construction | Tyvek with polymeric barrier coating |
Product Type | Sleeve |
Material | Tyvek |
Color | Yellow |
Seam Type | Stitched, Over-taped |
Cuff Style | Elastic |
Finish | Polymeric barrier coating |
Certifications/Compliance | EN 1149-5 |
Length (Metric) | 45 cm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tychem 2000 C Sleeve >
Spot an opportunity for improvement?Share a Content Correction